missing translation for 'onlineSavingsMsg'
Learn More

GFAP Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marke:  Invitrogen PA110019

Weitere Details : Netto-Gewicht : 0.01000kg

Artikelnummer. 11505872

  • 459.00 € / 100 Mikroliter
Lager vorrätig
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

By Western blot, two bands are detected in rat cerebellum homogenate: a prominent band at 50 kDa corresponding to rodent GFAP and a lower band at ∽45 kDa that most likely represents a proteolytic fragment derived from the GFAP molecule.

GFAP Polyclonal antibody specifically detects GFAP in Bovine, Equine, Human, Mouse, Porcine, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry, Western Blot.
Spezifikation

Spezifikation

GFAP
Polyclonal
Unconjugated
GFAP
6330404F12Rik; 65 kDa glutamic acid decarboxylase; AI836096; ALXDRD; Astrocyte or Intermediate Filament Protein; cb345; class III intermediate filament protein; etID36982.3; FLJ45472; GAD(65); Gad2; Gad-2; GAD65; GAD-65; GFAP; GFAP epsilon; gfap protein; gfapl; glial fibrillary acidic protein; Glial Fibrillary Acidic Protein (GFAP); glial fibrillary acidic protein alpha; glutamate decarboxylase 2; Glutamate decarboxylase 65 kDa isoform; glutamic acid decarboxylase 2; I79_011607; intermediate filament; intermediate filament protein; intermediate filament protein; GFAP; Unknown (protein for MGC:139638); wu:fb34h11; wu:fk42c12; xx:af506734; zgc:110485; zGFAP; intermediate filament protein; zrf-1 antigen
Rabbit
100 μL
Primary
Bovine, Equine, Human, Mouse, Porcine, Rat
Antibody
IgG
Immunocytochemistry, Immunohistochemistry, Western Blot
Conc. Not Determined
whole serum with 5mM sodium azide
P03995, P14136, P47819, Q28115
GFAP
Recombinant full length human GFAP isotype 1 expressed in and purified from E. coli.
RUO
100033970, 14580, 24387, 2670, 281189, 396562
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote