missing translation for 'onlineSavingsMsg'
Learn More

GFI1 Monoclonal Antibody (5D7), Invitrogen™

Mouse Monoclonal Antibody

Marke:  Invitrogen MA515688

Weitere Details : Netto-Gewicht : 0.01000kg

Artikelnummer. 11365653

  • 462.00 € / 100 Mikroliter
Verfügbar ab: 22-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

MA5-15688 targets GFI1 in indirect ELISA, WB applications and shows reactivity with Human samples. The MA5-15688 immunogen is purified recombinant fragment of human GFI1 expressed in E. Coli. MA5-15688 detects GFI1 which has a predicted molecular weight of approximately 45kDa.

GFI1 Monoclonal antibody specifically detects GFI1 in Human samples. It is validated for ELISA, Western Blot.
Spezifikation

Spezifikation

GFI1
Monoclonal
Conc. Not Determined
ascites with 0.03% sodium azide
Q99684
GFI1
Purified recombinant fragment of human GFI1 expressed in E. Coli.
RUO
2672
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
ELISA, Western Blot
5D7
Unconjugated
GFI1
AW495828; FLJ94509; Gfi1; GFI-1; GFI1A; growth factor independence-1; growth factor independent 1; growth factor independent 1 transcription repressor; growth factor independent 1 transcriptional repressor; growth factor independent protein 1; Growth factor independent-1; Pal1; Pal-1; SCN2; Zinc finger protein 163; zinc finger protein Gfi-1; ZNF163
Mouse
100 μL
Primary
Human
Antibody
IgG1
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote