missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GFR alpha-1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA112532
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
A suggested positive control for this product is human brain lysate.
GFRa-1 binds GDNF specifically and mediates activation of the Ret protein tyrosine kinase (PTK). Thus, GDNF, GFRa and the Ret PTK form a complex to transduce GDNF signal and to mediate GDNF function.Spezifikation
GFR alpha-1 | |
Polyclonal | |
PBS with 0.02% sodium azide | |
P56159, P97785, Q62997 | |
GFRA1 | |
Synthetic peptide corresponding to residues 369-382 of human GFR -1. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Immunohistochemistry, Western Blot | |
Unconjugated | |
GFRA1 | |
DKFZp313E1012, DKFZp686J0156, FLJ10561, FLJ31546, GDNFR, GDNFRA, GFR-ALPHA-1, MGC23045, RET1L, RETL1, TRNR1 | |
Rabbit | |
Ion-exchange chromatography | |
RUO | |
14585, 25454, 2674 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |