Learn More
GGA2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA529464
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: HepG2, Molt-4, Raji. Predicted reactivity: Mouse (94%), Rat (93%), Bovine (96%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
GGA2 Polyclonal antibody specifically detects GGA2 in Human samples. It is validated for Immunocytochemistry, Western Blot.Spezifikation
GGA2 | |
Polyclonal | |
Unconjugated | |
GGA2 | |
1200007E24Rik; ADP-ribosylation factor binding protein 2; ADP-ribosylation factor-binding protein GGA2; gam; Gamma-adaptin-related protein 2; Gga2; golgi associated, gamma adaptin ear containing, ARF binding protein 2; golgi-associated, gamma adaptin ear containing, ARF binding protein 2; golgi-localized, gamma ear-containing, ARF-binding protein 2; Kiaa1080; mKIAA1080; VEAR; VHS domain and ear domain of gamma-adaptin; VHS domain and ear domain-containing protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23062 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Western Blot | |
1 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
Q9UJY4 | |
GGA2 | |
Recombinant protein fragment corresponding to a region within amino acids 20 and 280 of Human GGA2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |