Learn More
Ghrelin Monoclonal Antibody (121), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen ABS05212102
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Specificity: ABS 052-121-02 cross-reacts with rat ghrelin, and is predicted to react with mouse, dog and cat based on sequence homology. Reactivity: ABS 052-121-02 binds human ghrelin in solution. ABS 052-121-02 can be used as a detection antibody in a sandwich ELISA with capture antibody ABS 050-45-02. NOTE: Concentration is lot-dependent and can vary from 0.85-1.15 mg/mL
Ghrelin Monoclonal antibody specifically detects Ghrelin in Canine, Feline, Human, Mouse, Rat samples. It is validated for ELISA, Western Blot.Spezifikation
Ghrelin | |
Monoclonal | |
0.9 mg/mL | |
PBS with 15mM sodium azide; pH 7.4 | |
Q6BEG6, Q9BEF8, Q9EQX0, Q9QYH7, Q9UBU3 | |
GHRL | |
Synthetic ghrelin (14-28). | |
400 μL | |
Primary | |
Canine, Feline, Human, Mouse, Rat | |
Antibody | |
IgG2b κ |
ELISA, Western Blot | |
121 | |
Unconjugated | |
GHRL | |
2210006E23Rik; AMCBX1; Appetite-regulating hormone; appetite-regulating hormone; uncharacterized protein LOC58991; exon 2- and 3-deleted preproghrelin; exon 2-deleted preproghrelin; FIP3; FIP-3; Fip3p; Ghr; Ghrelin; ghrelin 27; ghrelin 28; ghrelin and obestatin prepropeptide; ghrelin precursor; ghrelin, growth hormone secretagogue receptor ligand; ghrelin/obestatin; ghrelin/obestatin preprohormone; ghrelin/obestatin prepropeptide; Ghrelin-27; Ghrelin-28; ghrelin-like protein; GHRL; Growth hormone secretagogue; growth hormone-releasing peptide; IKK-gamma; In2c-preproghrelin; IP; IP1; IP2; IPD2; m46; Motilin-related peptide; MTLRP; MTLRPAP; NEMO; Obestatin; Obestatin-13; Obestatin-23; OTTHUMP00000207794; OTTHUMP00000207795; OTTHUMP00000207796; OTTHUMP00000207798; prepro-appetite regulatory hormone; preproghrelin; proghrelin; Protein M46; UNQ524/PRO1066 | |
Mouse | |
Protein A/G | |
RUO | |
403587, 493844, 51738, 58991, 59301 | |
4° C, store in dark | |
Liquid |