missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ghrelin Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marke: Invitrogen PA518409
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Ghrelin Polyclonal antibody specifically detects Ghrelin in Human, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).Spezifikation
Ghrelin | |
Polyclonal | |
Unconjugated | |
GHRL | |
2210006E23Rik; AMCBX1; Appetite-regulating hormone; appetite-regulating hormone; uncharacterized protein LOC58991; exon 2- and 3-deleted preproghrelin; exon 2-deleted preproghrelin; FIP3; FIP-3; Fip3p; Ghr; Ghrelin; ghrelin 27; ghrelin 28; ghrelin and obestatin prepropeptide; ghrelin precursor; ghrelin, growth hormone secretagogue receptor ligand; ghrelin/obestatin; ghrelin/obestatin preprohormone; ghrelin/obestatin prepropeptide; Ghrelin-27; Ghrelin-28; ghrelin-like protein; GHRL; Growth hormone secretagogue; growth hormone-releasing peptide; IKK-gamma; In2c-preproghrelin; IP; IP1; IP2; IPD2; m46; Motilin-related peptide; MTLRP; MTLRPAP; NEMO; Obestatin; Obestatin-13; Obestatin-23; OTTHUMP00000207794; OTTHUMP00000207795; OTTHUMP00000207796; OTTHUMP00000207798; prepro-appetite regulatory hormone; preproghrelin; proghrelin; Protein M46; UNQ524/PRO1066 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
51738, 59301 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q9QYH7, Q9UBU3 | |
GHRL | |
Synthetic peptide sequence (QDILWEEAKEAPADK) corresponding to the C-terminus amino acids of GHRL | |
100 μg | |
Primary | |
Human, Rat | |
Antibody | |
IgG |