Learn More
GHSR Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA528752
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: NT2D1, PC-3, U87-MG, SK-N-SH, mouse hypothalamus, mouse brain, rat brain. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
GHSR Polyclonal antibody specifically detects GHSR in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
GHSR | |
Polyclonal | |
Unconjugated | |
Ghsr | |
C530020I22Rik; GHDP; GH-releasing peptide receptor; Ghrelin receptor; GHRP; Ghrp receptor; GHS receptor; GHSR; GHS-R; G-protein coupled receptor; growth hormone secretagogue receptor; growth hormone secretagogue receptor type 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
208188, 2693, 84022 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
O08725, Q92847, Q99P50 | |
Ghsr | |
Synthetic peptide corresponding to a region within amino acids 1 and 45 of Human GHS-R1 | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |