missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Giardia lamblia 65 kDa Monoclonal Antibody (S1H), Invitrogen™
Mouse Monoclonal Antibody
Marke: Thermo Scientific MA518207
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Purity is >90%. This antibody is recommended as a capture antibody in ELISA applications.
Giardia lamblia 65 kDa Monoclonal antibody specifically detects Giardia lamblia 65 kDa in Protozoa samples. It is validated for ELISA, Immunocytochemistry.Spezifikation
Giardia lamblia 65 kDa | |
Monoclonal | |
100 μg/mL | |
PBS with 0.1% sodium azide; pH 7.2 | |
Mouse | |
Protein A | |
RUO | |
Protozoa | |
Antibody | |
IgG1 |
ELISA, Immunocytochemistry | |
S1H | |
Unconjugated | |
G. lamblia | |
Giardia lamblia native protein | |
100 μg | |
Primary | |
4° C | |
Liquid |