Learn More
Giardia lamblia 65 kDa Monoclonal Antibody (D33I), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA17440
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
The MA1-7440 antibody reacts with Giardia lamblia 65 kDa Ag in protozoan samples. MA1-7440 has been successfully used in ELISA, Immunofluorescence and Western blot applications. The MA1-7440 antibody was raised against Giardia 65 kDa antigen.
Giardia lamblia 65 kDa Monoclonal antibody specifically detects Giardia lamblia 65 kDa in Protozoa samples. It is validated for ELISA, Immunocytochemistry, Western Blot.Spezifikation
Giardia lamblia 65 kDa | |
Monoclonal | |
0.1 mg/mL | |
PBS with 0.1% sodium azide; pH 7.2 | |
Mouse | |
Protein A | |
RUO | |
Protozoa | |
Antibody | |
IgG2b |
ELISA, Immunocytochemistry, Western Blot | |
D33I | |
Unconjugated | |
G. lamblia | |
Giardia 65 kDa antigen. | |
100 μg | |
Primary | |
4° C | |
Liquid |