missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IFI30 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA521533
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: Raji. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
IFI30 Polyclonal antibody specifically detects IFI30 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Spezifikation
IFI30 | |
Polyclonal | |
Unconjugated | |
IFI30 | |
Gamma-interferon-inducible lysosomal thiol reductase; gamma-interferon-inducible protein IP-30; GILT; IFI30; IFI-30; IFI30, lysosomal thiol reductase; interferon gamma inducible protein 30; interferon gamma-inducible protein 30 preproprotein; interferon, gamma-inducible protein 30; IP30; IP-30; Legumaturain | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10437 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
P13284 | |
IFI30 | |
Recombinant protein fragment contain a sequence corresponding to a region within amino acids 14 and 250 of Human GILT | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |