missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GIMAP1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA543539
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Peptide sequence: MGGRKMATDE ENVYGLEENA QSRQESTRRL ILVGRTGAGK SATGNSILGQ Sequence homology: Human: 100%
GIMAP1 Polyclonal antibody specifically detects GIMAP1 in Human samples. It is validated for Western Blot.Spezifikation
GIMAP1 | |
Polyclonal | |
Unconjugated | |
GIMAP1 | |
GIMAP1; GTPase; GTPase IMAP family member 1; GTPase, IMAP family member 1; hIMAP1; Ian2; IAP38; imap; Imap1; Imap38; immune associated nucleotide family member; immune-associated nucleotide-binding protein 2; immune-associated protein 38; immunity associated protein 1; immunity-associated protein; immunity-associated protein 1; immunity-associated protein, 38 kDa | |
Rabbit | |
Affinity Chromatography | |
RUO | |
170575 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q8WWP7 | |
GIMAP1 | |
Synthetic peptide directed towards the N-terminal of human GIMAP1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |