missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GIMAP5 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA528872
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: HeLaS3. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
GIMAP5 Polyclonal antibody specifically detects GIMAP5 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Spezifikation
GIMAP5 | |
Polyclonal | |
Unconjugated | |
GIMAP5 | |
D630024P16; E230026N22Rik; Gimap5; GTPase IMAP family member 5; GTPase of the immunity-associated protein 5; GTPase, IMAP family member 5; HIMAP3; Ian4; IAN-4; Ian4l1; IAN5; IAN-5; Iddm1; IMAP3; immune associated nucleotide 4 like 1; immune-associated nucleotide-binding protein 5; immunity associated protein 3; immunity-associated nucleotide 4 protein; Immunity-associated nucleotide 4-like 1 protein; immunity-associated nucleotide 5 protein; Immunity-associated protein 3; inhibitor of radiation- and OA-induced apoptosis, Irod/Ian5; IROD; lymphopenia; lyp | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55340 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
Q96F15 | |
GIMAP5 | |
Synthetic peptide corresponding to a region within amino acids 188 and 280 of Human GIMAP5 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |