missing translation for 'onlineSavingsMsg'
Learn More

GIMAP8 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marke:  Invitrogen PA521969

Weitere Details : Netto-Gewicht : 0.01000kg

Artikelnummer. 13240179

  • 473.00 € / 100 Mikrogramm
Verfügbar ab: 22-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

Recommended positive controls: 293T, A431, H1299, HeLaS3, HepG2. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

GIMAP8 Polyclonal antibody specifically detects GIMAP8 in Human samples. It is validated for Western Blot.
Spezifikation

Spezifikation

GIMAP8
Polyclonal
Unconjugated
GIMAP8
GIMAP8; Gm457; GTPase IMAP family member 8; GTPase, IMAP family member 8; hIAN6; human immune associated nucleotide 6; IAN6; IAN9; IAN-9; Iant; IMAP8; Immune-associated nucleotide-binding protein 9; immunity-associated protein 8; mGIMAP8; Protein IanT
Rabbit
Antigen affinity chromatography
RUO
155038
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot
1 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Q8ND71
GIMAP8
Synthetic peptide corresponding to a region within amino acids 26 and 118 of GIMAP8 (Uniprot ID#Q8ND71)
100 μL
Primary
Human
Antibody
IgG
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote