missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GINS2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA542838
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Peptide sequence: VRQQEAHAKL DNLTLMEINT SGTFLTQALN HMYKLRTNLQ PLESTQSQDF Sequence homology: Cow: 79%; Dog: 93%; Horse: 86%; Human: 100%; Mouse: 93%; Rat: 86%
GINS2 Polyclonal antibody specifically detects GINS2 in Human samples. It is validated for Western Blot.Spezifikation
GINS2 | |
Polyclonal | |
Unconjugated | |
GINS2 | |
2210013I18Rik; 4833427B12Rik; AI323585; CGI-122; DC5; DNA replication complex GINS protein PSF2; GINS complex subunit 2; GINS complex subunit 2 (Psf2 homolog); Gins2; HSPC037; Pfs2; PSF2; RGD1311055 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
51659 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9Y248 | |
GINS2 | |
Synthetic peptide directed towards the C-terminal of human GINS2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |