Learn More
GIPC1 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marke:  Invitrogen PA19049
Weitere Details : Netto-Gewicht : 0.01000kg
Artikelnummer. 11533891
  • 473.00 € / 100 Mikrogramm
Verfügbar ab: 23-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Beschreibung
Beschreibung
This antibody is predicted to react with bovine, mouse and rat based on sequence homology.
GIPC1 Polyclonal antibody specifically detects GIPC1 in Human samples. It is validated for Western Blot.
Spezifikation
Spezifikation
GIPC1
Polyclonal
Unconjugated
GIPC1
C19orf3; GAIP C-terminus-interacting protein; Gipc; GIPC PDZ domain containing family member 1; GIPC PDZ domain containing family, member 1; Gipc1; GLUT1 C-terminal binding protein; GLUT1 C-terminal-binding protein; GLUT1CBP; Glut1CIP; Hs.6454; IGF-1 receptor interacting protein 1; IIP-1; neurophilin1-IP; NIP; PDZ domain-containing protein GIPC1; regulator of G-protein signaling 19 interacting protein 1; regulator of G-protein signalling 19 interacting protein 1; Rgs19; RGS19-interacting protein 1; RGS19IP1; RGS-GAIP interacting protein C-terminus; RGS-GAIP interacting protein GIPC; RGS-GAIP interacting protein, C-terminus; RGS-GAIP-interacting protein; semaF cytoplasmic domain associated protein; semaF cytoplasmic domain-associated protein 1; SEMCAP; Semcap1; SEMCAP-1; SYNECTIIN; SYNECTIN; Tax interacting protein, clone 2; tax interaction protein 2; TaxIP2; TIP-2
Goat
Antigen affinity chromatography
RUO
10755
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot
0.5 mg/mL
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
O14908
GIPC1
Synthetic peptide corresponding to the C terminus amino acids GAIGDAKVGRY.