Learn More
GK5 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA534732
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: 293T, A431, Molt-4, Raji. Predicted reactivity: Human (99%), Rhesus Monkey (96%), Bovine (84%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
GK5 Polyclonal antibody specifically detects GK5 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Spezifikation
GK5 | |
Polyclonal | |
Unconjugated | |
GK5 | |
ATP:glycerol 3-phosphotransferase 5; AV095337; C330018K18Rik; G630067D24Rik; GK 5; Gk5; glycerokinase 5; glycerol kinase 5 (putative); putative glycerol kinase 5; RGD1311223 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
256356 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.74 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
Q6ZS86 | |
GK5 | |
Recombinant protein fragment contains a sequence corresponding to a region within amino acids 9 and 297 of GK5 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |