Learn More
Rabbit anti-Goat IgG (H+L) Cross-Adsorbed Secondary Antibody, DyLight™ 488, Invitrogen™
Rabbit Polyclonal Secondary Antibody
Marke: Invitrogen SA510078
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
This antibody is cross-adsorbed and exhibits minimum reactivity to chicken, horse, human, mouse, pig and rat. For Western blot applications, 5% non-fat dry milk in PBST or TBST is recommended for blocking and incubation of antibodies. BSA is not recommended.
Goat IgG (H+L) Cross-Adsorbed Polyclonal antibody specifically detects Goat IgG (H+L) Cross-Adsorbed in Goat samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot.Spezifikation
Goat IgG (H+L) Cross-Adsorbed | |
Polyclonal | |
DyLight 488 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
Goat | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 6.8 to 7.4 | |
Goat IgG-heavy and light chain | |
500 μg | |
Secondary | |
4° C | |
IgG |