missing translation for 'onlineSavingsMsg'
Learn More
Learn More
F(ab')2-Rabbit anti-Goat IgG (H+L) Cross-Adsorbed Secondary Antibody, Invitrogen™
F(ab')2-Rabbit Polyclonal Secondary Antibody
Marke: Invitrogen A24454
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Goat IgG (H+L) Cross-Adsorbed Polyclonal antibody specifically detects Goat IgG (H+L) Cross-Adsorbed in Goat samples. It is validated for ELISA.Spezifikation
Goat IgG (H+L) Cross-Adsorbed | |
Polyclonal | |
Unconjugated | |
Rabbit | |
Affinity Chromatography | |
RUO | |
Goat | |
Liquid |
ELISA | |
> 4.5 mg/mL | |
PBS with 0.05% sodium azide; pH 7.2 | |
Gamma Immunoglobins Heavy and Light chains | |
1 mg | |
Secondary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
IgG |