Learn More
Gram Negative Endotoxin Monoclonal Antibody (GNE11-270.3.1), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA110688
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
MA1-10688 detects Endotoxin from K. pneumoniae, Sh. sonnei, and S. typhimurium samples. MA1-10688 has been successfully used in ELISA applications. The MA1-10688 immunogen is E. coli serotypes: O:111, B4 J5.
Gram Negative Endotoxin Monoclonal antibody specifically detects Gram Negative Endotoxin in Bacteria samples. It is validated for ELISA.Spezifikation
Gram Negative Endotoxin | |
Monoclonal | |
0.74 mg/mL | |
PBS with no preservative; pH 7.4 | |
E. coli serotypes: O:111, B4 J5. | |
500 μg | |
Primary | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA | |
GNE11-270.3.1 | |
Unconjugated | |
Mouse | |
Protein A | |
RUO | |
Bacteria | |
Antibody | |
IgG2a |