Learn More
IgG (H+L) Rabbit anti-Guinea Pig, AP, Invitrogen™
Rabbit Polyclonal Secondary Antibody
Marke:  Invitrogen A18886
Weitere Details : Netto-Gewicht : 0.01000kg
Artikelnummer. 15291688
Dieser Artikel ist im Moment nicht verfügbar oder wird nicht mehr hergestellt.
Schauen Sie bitte auf unsere Produktseite nach möglichen Alternativen
Alternative Produkte anzeigen »
Beschreibung
Beschreibung
The sensitivity of each lot of antibody is confirmed using ELISA. The specificity of each lot of antibody is confirmed by isoelectric focusing (IEF).
Anti-Guinea Pig secondary antibodies are affinity-purified antibodies with well-characterized specificity for guinea pig immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Spezifikation
Spezifikation
Guinea Pig IgG (H+L)
Polyclonal
Rabbit
Affinity chromatography
RUO
Guinea Pig
Liquid
ELISA, Immunohistochemistry, Western Blot
Alkaline Phosphatase (AP)
Gamma Immunoglobins Heavy and Light chains
500 μg
Secondary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.