Learn More
H-2Kb Mouse anti-Mouse, PE, Clone: AF6-88.5, BD
Mouse Monoclonal Antibody
Marke: BD Biosciences 561072
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
The AF6-88.5 antibody reacts with the H-2Kb MHC class I alloantigen. Reactivity with other haplotypes (e.g., d, f, j, k, p, q, r, s, u, v) has not been observed.
Spezifikation
H-2Kb | |
Monoclonal | |
0.2mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse C57BL splenocytes | |
25 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
AF6-88.5 | |
PE | |
Mouse | |
Affinity Purified | |
RUO | |
Mouse | |
IgG2a κ |
For Research Use Only.