Learn More
Rabbit anti-Syrian Hamster IgG (H+L) Cross-Adsorbed Secondary Antibody, HRP, Invitrogen™
Rabbit Polyclonal Secondary Antibody
Marke: Invitrogen A18895
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Rehydrate with 1.1 mL of deionized water and let stand 30 minutes at room temperature to dissolve. (Product has been overfilled to ensure complete recovery.) Centrifuge to remove any particulates. Prepare fresh working dilution daily. Store lyophilized material at 2-8 °C. For long term storage after reconstitution, dilute with 50% glycerol and store at -20 °C as a liquid. Based on Immunoelectrophoresis, no reactivity is observed to: non-immunoglobulin hamster serum immunoglobulins, human serum proteins.
Syrian Hamster IgG (H+L) Cross-Adsorbed Polyclonal antibody specifically detects Syrian Hamster IgG (H+L) Cross-Adsorbed in Hamster samples. It is validated for ELISA, Immunohistochemistry, Western Blot.Spezifikation
Syrian Hamster IgG (H+L) Cross-Adsorbed | |
Polyclonal | |
HRP | |
Rabbit | |
Affinity chromatography | |
RUO | |
Hamster | |
Lyophilized |
ELISA, Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 1% BSA and 0.1% ProClin 150; pH 7.2 | |
Gamma Immunoglobins Heavy and Light chains | |
1 mg | |
Secondary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
IgG |