Learn More
HSC70 Monoclonal Antibody (N27F3-4), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA110890
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
MA1-10890 detects Heat Shock Protein 70/Heat Shock Cognate Protein 70 in human, mouse, rat, rabbit, monkey, guinea pig, hamster, chicken, bovine, canine, ovine, porcine, fish, Xenopus, Drosophila, C. elegans, and plant samples. MA1-10890 has been successfully used in Western blot, immunohistochemistry, immunoprecipitation and flow cytometry procedures. By Western blot, MA1-10890 is specific for the ∽72 kDa Heat Shock Protein 70/Heat Shock Cognate Protein 70 protein. Positive control of HeLa cell lysate recommended. MA1-10890 immunogen is purified recombinant Hsp70/Hsc70.
HSC70 Monoclonal antibody specifically detects HSC70 in Human, Many, Mouse, Rat samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot.Spezifikation
HSC70 | |
Monoclonal | |
1.0 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.4 | |
P11142, P63017, P63018 | |
Hspa8 | |
Purified recombinant Hsp70/Hsc70. | |
100 μg | |
Primary | |
Human, Multi-species, Mouse, Rat | |
Antibody | |
IgG1 |
Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot | |
N27F3-4 | |
Unconjugated | |
Hspa8 | |
2410008N15Rik; AAG2; constitutive heat shock protein 70; constitutively expressed heat shock protein; epididymis luminal protein 33; epididymis secretory sperm binding protein Li 72p; FAM10A1; Heat shock 70 kDa protein 8; heat shock 70kd protein 10; heat shock 70kd protein 10 (HSC71); heat shock 70kD protein 8; heat shock 70kDa protein 8; heat shock cognate; heat shock cognate 70; heat shock cognate 71 kD protein; heat shock cognate 71 kDa protein; heat shock cognate 71 kDa protein-like protein; heat shock cognate 71-kd protein; heat shock cognate hsc73; heat shock cognate protein 54; Heat shock cognate protein 70; heat shock protein 70 cognate; heat shock protein 8; heat shock protein A8; heat shock protein cognate 70; heat shock protein family A (Hsp70) member 8; HEL-33; HEL-S-72p; HIP; HSC54; Hsc70; Hsc70 ATPase; HSC70 protein; Hsc71; Hsc73; HSP70; HSP71; HSP73; Hspa10; Hspa8; hspa8 protein; I79_022974; LAP; LAP1; LAP-1; lipopolysaccharide-associated protein 1; LPS-associated protei | |
Mouse | |
Protein G | |
RUO | |
15481, 24468, 3312 | |
4° C | |
Liquid |