missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSV Type 1 VP21/VP22a Monoclonal Antibody (LP13), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA516284
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
HSV Type 1 VP21/VP22a Monoclonal antibody specifically detects HSV Type 1 VP21/VP22a in Virus samples. It is validated for Immunocytochemistry.Spezifikation
HSV Type 1 VP21/VP22a | |
Monoclonal | |
6.64 mg/mL | |
PBS with 15mM sodium azide; pH 7.5 | |
Mouse | |
Protein A | |
RUO | |
Virus | |
Antibody | |
IgG2a |
Immunocytochemistry | |
LP13 | |
Unconjugated | |
Herpes Simplex 1; Herpes Simplex I; HHV; HSV; HSV Type 1; HSV1; KITH_HHV1S | |
HSV I (strain HFEM) | |
2 mg | |
Primary | |
4° C | |
Liquid |