missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSV Type 2 Monoclonal Antibody (AP1), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA516286
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
HSV Type 2 Monoclonal antibody specifically detects HSV Type 2 in Virus samples. It is validated for Immunocytochemistry.Spezifikation
HSV Type 2 | |
Monoclonal | |
1 mg/mL | |
PBS with 15mM sodium azide; pH 7.5 | |
Mouse | |
Protein A | |
RUO | |
Virus | |
Antibody | |
IgG2a |
Immunocytochemistry | |
AP1 | |
Unconjugated | |
Herpes Simplex 2; Herpes Simplex II; herpes simplex virus 2; HHV-2; HSV Type 2; HSV Type II; HSV2; HSV-2 | |
HSV II (strain 25766) | |
2 mg | |
Primary | |
4° C | |
Liquid |