missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSV Type 1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA538569
Weitere Details : Netto-Gewicht : 0.00900kg
Beschreibung
HSV is a member of the herpes virus family (Herpesviridae), Herpes simplex virus types 1 and 2 (HSV-1 and HSV-2), are ubiquitously contagious to humans. Also known as Human herpes virus 1 and 2 (HHV-1 and -2), respectively, HSV-1 strains produce cold sores, while HSV-2 produce most genital herpes. They can be spread when an infected person is producing and shedding the virus. The virion form of herpes viruses consists of a relatively large double-stranded, linear DNA genome encased within the capsid and wrapped in the viral envelope. The different HSV strains carry over 70 open reading frames (ORFs). HSV has been linked to Alzheimer′s disease.Spezifikation
HSV Type 1 | |
Polyclonal | |
Unconjugated | |
HSV1; Herpes Simplex I; HSV Type 1; Herpes Simplex 1; HHV; HSV; Herpes Simplex Type 1; KITH_HHV1S | |
A synthetic peptide derived from the internal region of human KITH_HHV1S | |
100 μg | |
Primary | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
Virus | |
Antibody | |
IgG |