missing translation for 'onlineSavingsMsg'
Learn More

HGF Monoclonal Antibody (24612.111), Invitrogen™

Mouse Monoclonal Antibody

Marke:  Invitrogen MA124767

Weitere Details : Netto-Gewicht : 0.01000kg

Artikelnummer. 11524792

  • 470.00 € / 100 Mikrogramm
Verfügbar ab: 23-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

Monoclonal Anti-Human HGF was tested for its ability to neutralize the bioactivity of rhHGF in a cell proliferation assay using 4MBr-5 cells, a monkey epithelial cell line responsive to HGF. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

HGF Monoclonal antibody specifically detects HGF in Human samples. It is validated for ELISA, Inhibition Assays, Neutralization, Western Blot.
Spezifikation

Spezifikation

HGF
Monoclonal
0.5 mg/mL
PBS with 5% trehalose and no preservative
P14210
Hgf
Recombinant human hepatocyte growth factor, expressed in the insect Sf 21 cells.
100 μg
Primary
Human
Antibody
IgG1
ELISA, Inhibition Assays, Neutralization, Western Blot
24612.111
Unconjugated
Hgf
C230052L06Rik; DFNB39; Fibroblast derived tumor cytotoxic factor; fibroblast-derived tumor cytotoxic factor; F-TCF; hepatocyte growth factor; Hepatocyte growth factor (hepapoietin A; hepatocyte growth factor (hepapoietin A; scatter factor); Hepatocyte growth factor alpha chain; Hepatocyte growth factor beta chain; Hepatopoeitin A; hepatopoeitin-A; HepatopoietinA; hepatopoietin-A; Hgf; HGF/SF; HGFB; HPTA; Lung fibroblast derived mitogen; lung fibroblast-derived mitogen; NK1; NK2; Scatter factor; scatter factor); SF; SF/HGF
Mouse
Protein A
RUO
3082
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote