missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HGF Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marke: Invitrogen PA128024
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
HGF Polyclonal antibody specifically detects HGF in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Spezifikation
HGF | |
Polyclonal | |
Unconjugated | |
Hgf | |
C230052L06Rik; DFNB39; Fibroblast derived tumor cytotoxic factor; fibroblast-derived tumor cytotoxic factor; F-TCF; hepatocyte growth factor; Hepatocyte growth factor (hepapoietin A; hepatocyte growth factor (hepapoietin A; scatter factor); Hepatocyte growth factor alpha chain; Hepatocyte growth factor beta chain; Hepatopoeitin A; hepatopoeitin-A; HepatopoietinA; hepatopoietin-A; Hgf; HGF/SF; HGFB; HPTA; Lung fibroblast derived mitogen; lung fibroblast-derived mitogen; NK1; NK2; Scatter factor; scatter factor); SF; SF/HGF | |
Goat | |
Antigen affinity chromatography | |
RUO | |
3082 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.1 mg/mL | |
PBS with 5% trehalose and no preservative; pH 7.4 | |
P14210 | |
Hgf | |
Recombinant, human HGF (rhHGF) expressed in SF 21 | |
25 μg | |
Primary | |
Human | |
Antibody | |
IgG |