Learn More
HGS Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA527491
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: H1299, HeLa, mouse brain, rat brain. Predicted reactivity: Mouse (85%), Rat (82%), Dog (92%), Rhesus Monkey (98%), Bovine (89%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
HGS Polyclonal antibody specifically detects HGS in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
HGS | |
Polyclonal | |
Unconjugated | |
HGS | |
hepatocyte growth factor-regulated tyrosine kinase substrate; HGF-regulated tyrosine kinase substrate; HGNC:4897; Hgr; Hgs; Hrs; Hrs2; human growth factor-regulated tyrosine kinase substrate; protein pp110; SNAP-25-interacting protein Hrs-2; ZFYVE8 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
15239, 56084, 9146 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
O14964, Q99LI8, Q9JJ50 | |
HGS | |
Recombinant fragment corresponding to a region within amino acids 611 and 777 of Human HGS | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |