Learn More
HHIP Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA529529
Additional Details : Netto-Gewicht : 0.01000kg
Description
Recommended positive controls: NT2D1. Predicted reactivity: Mouse (97%), Rat (96%), Rhesus Monkey (99%), Bovine (96%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
HHIP Polyclonal antibody specifically detects HHIP in Human samples. It is validated for Western Blot.Specifications
HHIP | |
Polyclonal | |
Unconjugated | |
HHIP | |
hedgehog interacting protein; Hedgehog-interacting protein; Hedgehog-interacting protein-like; Hhip; Hhip1; HIP; RGD1564108; UNQ5825/PRO19644 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
64399 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.89 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q96QV1 | |
HHIP | |
Recombinant fragment corresponding to a region within amino acids 386 and 635 of Human HHIP | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |