Learn More
HIBADH Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA532071
Additional Details : Netto-Gewicht : 0.01000kg
Description
Recommended positive controls: A549, GL261. Predicted reactivity: Mouse (94%), Rat (93%), Xenopus laevis (91%), Chicken (88%), Bovine (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
HIBADH Polyclonal antibody specifically detects HIBADH in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
HIBADH | |
Polyclonal | |
Unconjugated | |
HIBADH | |
3'-hydroxyisobutyrate dehydrogenase; 3-hydroxyisobutyrate dehydrogenase; 3-hydroxyisobutyrate dehydrogenase, mitochondrial; 6430402H10Rik; AI265272; Hibadh; NS5ATP1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
11112, 58875 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
P31937, Q99L13 | |
HIBADH | |
Recombinant fragment corresponding to a region within amino acids 128 and 336 of Human HIBADH | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |