Learn More
HIBCH Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA528949
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: 293T, HepG2, Molt-4. Predicted reactivity: Mouse (81%), Bovine (84%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
HIBCH Polyclonal antibody specifically detects HIBCH in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
HIBCH | |
Polyclonal | |
Unconjugated | |
HIBCH | |
2610509I15Rik; 3-hydroxyisobutyryl-CoA hydrolase; 3-hydroxyisobutyryl-CoA hydrolase, mitochondrial; 3-hydroxyisobutyryl-coenzyme A hydrolase; AI648812; Hibch; HIB-CoA hydrolase; HIBYLCOAH; HIBYL-CoA-H; testicular tissue protein Li 86 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
26275 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
Q6NVY1 | |
HIBCH | |
Full length human HIBCH Recombinant protein. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |