missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HIC2 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marke: Invitrogen PA537931
Weitere Details : Netto-Gewicht : 0.00900kg
Beschreibung
This antibody is predicted to react with mouse, rat, dog, pig and cow based on sequence homology.
HIC2 Polyclonal antibody specifically detects HIC2 in Human samples. It is validated for Western Blot.Spezifikation
HIC2 | |
Polyclonal | |
Unconjugated | |
HIC2 | |
AA409108; HIC ZBTB transcriptional repressor 2; HIC1-related gene on chromosome 22 protein; HIC2; hic-2; Hic-3; HRG22; hypermethylated in cancer 2; hypermethylated in cancer 2 protein; Kiaa1020; LOW QUALITY PROTEIN: hypermethylated in cancer 2 protein; mKIAA1020; ZBTB30; Zinc finger and BTB domain-containing protein 30; ZNF907 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
23119 | |
-20°C | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q96JB3 | |
HIC2 | |
Peptide with sequence C-PQELPQAKGSDDE, from the internal region of HIC2 (aa188-200) | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |