missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HIC5 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA517540
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
It is not recommended to aliquot this antibody.
HIC5 Polyclonal antibody specifically detects HIC5 in Bovine, Human, Non-human Primate samples. It is validated for Immunoprecipitation, Western Blot.Spezifikation
HIC5 | |
Polyclonal | |
0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and no preservative; pH 7.5 | |
O43294, Q3MHZ4 | |
Tgfb1i1 | |
Synthetic peptide corresponding to residues surrounding Ala104 of human Hic-5 | |
100 μL | |
Primary | |
Bovine, Human, Non-human Primate | |
Antibody | |
IgG |
Immunoprecipitation, Western Blot | |
Unconjugated | |
Tgfb1i1 | |
Androgen receptor coactivator 55 kDa protein; androgen receptor coactivator ARA55; androgen receptor-associated protein of 55 kDa; Ara55; Hic5; hic-5; hydrogen peroxide-inducible clone 5 protein; hydrogen peroxide-inducible clone-5; TGF beta-stimulated clone 5; Tgfb1i1; transforming growth factor beta 1 induced transcript 1; transforming growth factor beta-1-induced transcript 1 protein; TSC-5 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
515834, 7041 | |
-20°C | |
Liquid |