missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HIC2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA537293
Weitere Details : Netto-Gewicht : 0.00910kg
Beschreibung
This antibody detects endogenous protein at a molecular weight of 72 kDa. Purity is >95% by SDS-PAGE.
HIC2 Polyclonal antibody specifically detects HIC2 in Human, Mouse samples. It is validated for Western Blot.Spezifikation
HIC2 | |
Polyclonal | |
Unconjugated | |
HIC2 | |
AA409108; HIC ZBTB transcriptional repressor 2; HIC1-related gene on chromosome 22 protein; HIC2; hic-2; Hic-3; HRG22; hypermethylated in cancer 2; hypermethylated in cancer 2 protein; Kiaa1020; LOW QUALITY PROTEIN: hypermethylated in cancer 2 protein; mKIAA1020; ZBTB30; Zinc finger and BTB domain-containing protein 30; ZNF907 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23119, 58180 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96JB3, Q9JLZ6 | |
HIC2 | |
A synthetic peptide corresponding to residues in human HIC2 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |