Learn More
HICE1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA545046
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Peptide sequence: QTRGKMSEGG RKSSLLQKSK ADSSGVGKGD LQSTLLEGHG TAPPDLDLSA Sequence homology: Human: 100%; Pig: 75%
HICE1 Polyclonal antibody specifically detects HICE1 in Human samples. It is validated for Western Blot.Spezifikation
HICE1 | |
Polyclonal | |
Unconjugated | |
HAUS8 | |
2410004L22Rik; 4HAUS augmin-like complex, subunit 8; DGT4; HAUS augmin like complex subunit 8; HAUS augmin-like complex subunit 8; HAUS augmin-like complex, subunit 8; HAUS8; HEC1/NDC80 interacting, centrosome associated 1; HEC1/NDC80-interacting centrosome-associated protein 1; Hec1-interacting and centrosome-associated 1; Hice1; MGC20533; NY SAR 48; NYSAR48; NY-SAR-48; Sarcoma antigen NY-SAR-48; sarcoma antigen NY-SAR-48 homolog | |
Rabbit | |
Affinity Chromatography | |
RUO | |
93323 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9BT25 | |
HAUS8 | |
Synthetic peptide directed towards the N-terminal of human HAUS8 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |