missing translation for 'onlineSavingsMsg'
Learn More

HICE1 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marke:  Invitrogen PA521331

Weitere Details : Netto-Gewicht : 0.01000kg

Artikelnummer. 13209268

  • 475.00 € / 100 Mikroliter
Verfügbar ab: 19-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

Recommended positive controls: K562, HeLa. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

HICE1 Polyclonal antibody specifically detects HICE1 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.
Spezifikation

Spezifikation

HICE1
Polyclonal
Unconjugated
HAUS8
2410004L22Rik; 4HAUS augmin-like complex, subunit 8; DGT4; HAUS augmin like complex subunit 8; HAUS augmin-like complex subunit 8; HAUS augmin-like complex, subunit 8; HAUS8; HEC1/NDC80 interacting, centrosome associated 1; HEC1/NDC80-interacting centrosome-associated protein 1; Hec1-interacting and centrosome-associated 1; Hice1; MGC20533; NY SAR 48; NYSAR48; NY-SAR-48; Sarcoma antigen NY-SAR-48; sarcoma antigen NY-SAR-48 homolog
Rabbit
Antigen affinity chromatography
RUO
93323
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.17 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Q9BT25
HAUS8
Recombinant protein fragment contain a sequence corresponding to a region within amino acids 166 and 360 (Q9BT25) of HICE1
100 μL
Primary
Human
Antibody
IgG
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote