Learn More
HIF1A Monoclonal Antibody (H1alpha67), DyLight™ 488, Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA516009
Weitere Details : Netto-Gewicht : 0.10000kg
Beschreibung
In Western blot, multiple bands may be seen at 100-120 kDa representing post-translational modification of HIF-1 alpha. For WB, testing on nuclear extracts is recommended. Suggested positive control: Cos-7 nuclear extracts.
HIF1A Monoclonal antibody specifically detects HIF1A in Bovine, Canine, Feline, Ferret, Human, Mouse, Non-human Primate, Ovine, Porcine, Rabbit, Rat, X. laevis samples. It is validated for ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot.Spezifikation
HIF1A | |
Monoclonal | |
0.7 mg/mL | |
50mM sodium borate with 0.05% sodium azide | |
O35800, Q16665, Q61221, Q9I8A9, Q9XTA5 | |
hif-1a, HIF1A, hif1a.L | |
Fusion protein containing amino acids 432-528 of human HIF-1alpha. | |
100 μL | |
Primary | |
Bovine, Canine, Feline, Canine, Human, Mouse, Human, Ovine, Non-human Primate, Ovine, Porcine, Rabbit | |
Antibody | |
IgG2b |
ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Western Blot | |
H1alpha67 | |
DyLight 488 | |
HIF1A | |
AA959795; ARNT interacting protein; ARNT2; ARNT-interacting protein; Basic-helix-loop-helix-PAS protein MOP1; bHLHe78; Class E basic helix-loop-helix protein 78; HIF; hif 1; hif 1a; HIF1; HIF1 alpha; HIF-1 alpha; HIF1a; hif-1a; hif1a.L; HIF1alpha; HIF-1alpha; HIF-1-alpha; HIF1-alpha; HIF1-alpha protein; hypoxia inducible factor 1 alpha; hypoxia inducible factor 1 alpha subunit; hypoxia inducible factor 1 alpha subunit L homeolog; hypoxia inducible factor 1 subunit alpha; hypoxia inducible factor 1 subunit alpha L homeolog; hypoxia inducible factor 1, alpha subunit; hypoxia inducible factor 1, alpha subunit (basic helix-loop-helix transcription factor); hypoxia inducible factor-1 alpha; hypoxia-inducible factor 1 alpha; hypoxia-inducible factor 1 alpha isoform I.3; hypoxia-inducible factor 1 alpha subunit; hypoxia-inducible factor 1, alpha subunit (basic helix-loop-helix transcription factor); hypoxia-inducible factor 1, alpha subunit (basic helix-loop-helix transcription factor) L home | |
Mouse | |
Protein G | |
RUO | |
100009579, 100337573, 101083686, 15251, 281814, 29560, 3091, 380141, 396696, 443519, 445838, 480348 | |
4° C, store in dark | |
Liquid |