Learn More
HIF1A Monoclonal Antibody (H1alpha67), DyLight™ 550, Invitrogen™
Mouse Monoclonal Antibody
Marke:  Invitrogen MA516010
Weitere Details : Netto-Gewicht : 0.10000kg
Artikelnummer. 13237618
  • 508.00 € / 250 Mikroliter
Verfügbar ab: 19-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Beschreibung
Beschreibung
In Western blot, multiple bands may be seen at 100-120 kDa representing post-translational modification of HIF-1 alpha. For WB, testing on nuclear extracts is recommended. Suggested positive control: Cos-7 nuclear extracts.
HIF1A Monoclonal antibody specifically detects HIF1A in Bovine, Canine, Feline, Ferret, Human, Mouse, Non-human Primate, Ovine, Porcine, Rabbit, Rat, X. laevis samples. It is validated for ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot.
Spezifikation
Spezifikation
HIF1A
Monoclonal
0.90 mg/mL
50mM sodium borate with 0.05% sodium azide
O35800, Q16665, Q61221, Q9I8A9, Q9XTA5
hif-1a, HIF1A, hif1a.L
Fusion protein containing amino acids 432-528 of human HIF-1alpha.
100 μL
Primary
Bovine, Canine, Feline, Canine, Human, Mouse, Human, Mouse, Porcine, Ovine, Porcine, Rabbit
Antibody
IgG2b
ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
H1alpha67
DyLight 550
HIF1A
AA959795; ARNT interacting protein; ARNT2; ARNT-interacting protein; Basic-helix-loop-helix-PAS protein MOP1; bHLHe78; Class E basic helix-loop-helix protein 78; HIF; hif 1; hif 1a; HIF1; HIF1 alpha; HIF-1 alpha; HIF1a; hif-1a; hif1a.L; HIF1alpha; HIF-1alpha; HIF-1-alpha; HIF1-alpha; HIF1-alpha protein; hypoxia inducible factor 1 alpha; hypoxia inducible factor 1 alpha subunit; hypoxia inducible factor 1 alpha subunit L homeolog; hypoxia inducible factor 1 subunit alpha; hypoxia inducible factor 1 subunit alpha L homeolog; hypoxia inducible factor 1, alpha subunit; hypoxia inducible factor 1, alpha subunit (basic helix-loop-helix transcription factor); hypoxia inducible factor-1 alpha; hypoxia-inducible factor 1 alpha; hypoxia-inducible factor 1 alpha isoform I.3; hypoxia-inducible factor 1 alpha subunit; hypoxia-inducible factor 1, alpha subunit (basic helix-loop-helix transcription factor); hypoxia-inducible factor 1, alpha subunit (basic helix-loop-helix transcription factor) L home
Mouse
Protein G
RUO
100009579, 100337573, 101083686, 15251, 281814, 29560, 3091, 380141, 396696, 443519, 445838, 480348
4° C, store in dark