missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HIF1A Polyclonal Antibody, DyLight™ 650, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA522777
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
This antibody detects upregulation of HIF-1 alpha in hypoxic samples.
HIF1A Polyclonal antibody specifically detects HIF1A in Canine, Fish, Goat, Hamster, Human, Mouse, Non-human Primate, Porcine, Rabbit, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot.Spezifikation
HIF1A | |
Polyclonal | |
DyLight 650 | |
HIF1A | |
AA959795; ARNT interacting protein; ARNT2; ARNT-interacting protein; Basic-helix-loop-helix-PAS protein MOP1; bHLHe78; Class E basic helix-loop-helix protein 78; HIF; hif 1; hif 1a; HIF1; HIF1 alpha; HIF-1 alpha; HIF1a; hif-1a; hif1a.L; HIF1alpha; HIF-1alpha; HIF-1-alpha; HIF1-alpha; HIF1-alpha protein; hypoxia inducible factor 1 alpha; hypoxia inducible factor 1 alpha subunit; hypoxia inducible factor 1 alpha subunit L homeolog; hypoxia inducible factor 1 subunit alpha; hypoxia inducible factor 1 subunit alpha L homeolog; hypoxia inducible factor 1, alpha subunit; hypoxia inducible factor 1, alpha subunit (basic helix-loop-helix transcription factor); hypoxia inducible factor-1 alpha; hypoxia-inducible factor 1 alpha; hypoxia-inducible factor 1 alpha isoform I.3; hypoxia-inducible factor 1 alpha subunit; hypoxia-inducible factor 1, alpha subunit (basic helix-loop-helix transcription factor); hypoxia-inducible factor 1, alpha subunit (basic helix-loop-helix transcription factor) L home | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
100009579, 100751293, 100861391, 15251, 29560, 3091, 396696, 480348 | |
4° C, store in dark | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
50mM sodium borate with no preservative | |
O35800, Q16665, Q61221 | |
HIF1A | |
A fusion protein including residues 530-825 of the mouse HIF-1 alpha protein. | |
100 μL | |
Primary | |
Canine, Fish, Goat, Hamster, Human, Mouse, Non-human Primate, Porcine, Rabbit, Rat | |
Antibody | |
IgG |