missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HIF1A Monoclonal Antibody (ESEE122), HRP, Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA516019
Weitere Details : Netto-Gewicht : 0.10000kg
Beschreibung
Suggested positive control: Cos7 CoCl2-treated nuclear extract.
HIF1A Monoclonal antibody specifically detects HIF1A in Bovine, Canine, Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot.Spezifikation
HIF1A | |
Monoclonal | |
0.8 mg/mL | |
PBS with no preservative | |
O35800, Q16665, Q61221, Q9XTA5 | |
HIF1A | |
Human HIF-1 alpha amino acids 329-530. | |
100 μL | |
Primary | |
Bovine, Canine, Human, Mouse, Rat | |
Antibody | |
IgG1 |
Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot | |
ESEE122 | |
HRP | |
HIF1A | |
AA959795; ARNT interacting protein; ARNT2; ARNT-interacting protein; Basic-helix-loop-helix-PAS protein MOP1; bHLHe78; Class E basic helix-loop-helix protein 78; HIF; hif 1; hif 1a; HIF1; HIF1 alpha; HIF-1 alpha; HIF1a; hif-1a; hif1a.L; HIF1alpha; HIF-1alpha; HIF-1-alpha; HIF1-alpha; HIF1-alpha protein; hypoxia inducible factor 1 alpha; hypoxia inducible factor 1 alpha subunit; hypoxia inducible factor 1 alpha subunit L homeolog; hypoxia inducible factor 1 subunit alpha; hypoxia inducible factor 1 subunit alpha L homeolog; hypoxia inducible factor 1, alpha subunit; hypoxia inducible factor 1, alpha subunit (basic helix-loop-helix transcription factor); hypoxia inducible factor-1 alpha; hypoxia-inducible factor 1 alpha; hypoxia-inducible factor 1 alpha isoform I.3; hypoxia-inducible factor 1 alpha subunit; hypoxia-inducible factor 1, alpha subunit (basic helix-loop-helix transcription factor); hypoxia-inducible factor 1, alpha subunit (basic helix-loop-helix transcription factor) L home | |
Mouse | |
Protein G | |
RUO | |
15251, 281814, 29560, 3091, 480348 | |
4° C, store in dark | |
Liquid |