missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HIF-1 beta Monoclonal Antibody (H1beta234), DyLight™ 488, Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA516013
Weitere Details : Netto-Gewicht : 0.10000kg
Beschreibung
HIF-1 beta Monoclonal antibody specifically detects HIF-1 beta in Bovine, Ferret, Human, Mouse, Non-human Primate, Ovine, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
HIF-1 beta | |
Monoclonal | |
1.4 mg/mL | |
50mM sodium borate with no preservative | |
P27540, P41739, P53762, Q9BE97 | |
ARNT | |
Fusion protein containing amino acids 496-789 of human HIF-1 beta. | |
100 μL | |
Primary | |
Bovine, Ferret, Human, Mouse, Non-human Primate, Ovine, Rat | |
Antibody | |
IgG1 κ |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
H1beta234 | |
DyLight 488 | |
ARNT | |
AHA-1; ARNT; ARNT protein; Arnt1; ARNT1a; ARNT1b; aryl hydrocarbon receptor nuclear translocater; Aryl hydrocarbon receptor nuclear translocator; Aryl hydrocarbon receptor nuclear translocator 1; aryl hydrocarbon receptor nuclear translocator type 1a; aryl hydrocarbon receptor nuclear translocator type 1b; bHLHe2; Class E basic helix-loop-helix protein 2; D3Ertd557e; dioxin receptor, nuclear translocator; Drnt; ESTM42; hif 1; HIF1 beta; HIF-1 beta; Hif1b; HIF1BETA; HIF-1beta; HIF-1-beta; HIF1-beta; hypoxia-inducible factor 1 beta; hypoxia-inducible factor 1, beta subunit; hypoxia-inducible factor 1-beta; mKIAA4051; TANGO; W08714; zgc:136664 | |
Mouse | |
Protein G | |
RUO | |
11863, 25242, 281010, 405, 443285 | |
4° C, store in dark | |
Liquid |