Learn More
HIF-1 beta Monoclonal Antibody (H1beta234), DyLight™ 650, Invitrogen™
Mouse Monoclonal Antibody
Marke:  Invitrogen MA516012
Weitere Details : Netto-Gewicht : 0.10000kg
Artikelnummer. 13257618
  • 514.00 € / 100 Mikroliter
Verfügbar ab: 19-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Beschreibung
Beschreibung
HIF-1 beta Monoclonal antibody specifically detects HIF-1 beta in Bovine, Ferret, Human, Mouse, Non-human Primate, Ovine, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.
Spezifikation
Spezifikation
HIF-1 beta
Monoclonal
1.4 mg/mL
50mM sodium borate with no preservative
P27540, P41739, P53762, Q9BE97
ARNT
Fusion protein containing amino acids 496-789 of human HIF-1 beta.
100 μL
Primary
Bovine, Ferret, Human, Mouse, Non-human Primate, Ovine, Rat
Antibody
IgG1 κ
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
H1beta234
DyLight 650
ARNT
AHA-1; ARNT; ARNT protein; Arnt1; ARNT1a; ARNT1b; aryl hydrocarbon receptor nuclear translocater; Aryl hydrocarbon receptor nuclear translocator; Aryl hydrocarbon receptor nuclear translocator 1; aryl hydrocarbon receptor nuclear translocator type 1a; aryl hydrocarbon receptor nuclear translocator type 1b; bHLHe2; Class E basic helix-loop-helix protein 2; D3Ertd557e; dioxin receptor, nuclear translocator; Drnt; ESTM42; hif 1; HIF1 beta; HIF-1 beta; Hif1b; HIF1BETA; HIF-1beta; HIF-1-beta; HIF1-beta; hypoxia-inducible factor 1 beta; hypoxia-inducible factor 1, beta subunit; hypoxia-inducible factor 1-beta; mKIAA4051; TANGO; W08714; zgc:136664
Mouse
Protein G
RUO
11863, 25242, 281010, 405, 443285
4° C, store in dark