missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HIF-2 alpha Polyclonal Antibody, HRP, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA522696
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Suggested positive control: hypoxic lysates.
HIF-2 alpha Polyclonal antibody specifically detects HIF-2 alpha in Fish, Hamster, Human, Mouse, Non-human Primate, Ovine, Rabbit, Rat, Reptile samples. It is validated for ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot.Spezifikation
HIF-2 alpha | |
Polyclonal | |
HRP | |
EPAS1 | |
basic-helix-loop-helix-PAS protein MOP2; bHLHe73; Class E basic helix-loop-helix protein 73; ECYT4; endothelial PAS domain protein 1; endothelial PAS domain protein 1b; endothelial PAS domain-containing protein 1; endothelial PAS domain-containing protein 1b; EPAS; Epas1; EPAS-1; epas1b; Hif like protein; HIF1 alpha-like factor; HIF-1-alpha-like factor; HIF-1alpha-like factor; HIF1alpha-like factor; HIF2 alpha; HIF-2 alpha; HIF2A; hif-2a; hif-2ab; HIF-2alpha; HIF-2-alpha; HIF2-alpha; HIF-related factor; HLF; HLF (HIF1alpha-like factor); HRF; hypoxia inducible factor 2 alpha; hypoxia inducible factor 2, alpha subunit; hypoxia inducible factor 2a; hypoxia inducible transcription factor 2alpha; hypoxia-inducible factor 2 alpha; hypoxia-inducible factor 2-alpha; Member of PAS protein 2; mHLF; MOP2; PAS domain-containing protein 2; PASD2; wu:fq36f01 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
100348809, 100771160, 13819, 2034, 29452, 443286 | |
4° C, store in dark | |
Liquid |
ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot | |
1.03 mg/mL | |
PBS with no preservative | |
P97481, Q99814, Q9JHS1 | |
EPAS1 | |
A peptide derived from the C-terminus of mouse/human HIF-2 alpha protein. | |
100 μL | |
Primary | |
Fish, Hamster, Human, Mouse, Non-human Primate, Ovine, Rabbit, Rat, Reptile | |
Antibody | |
IgG |