Learn More
HIF3A Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Marke:  Invitrogen PA146117
Weitere Details : Netto-Gewicht : 0.01000kg
Artikelnummer. 11568940
Dieser Artikel ist im Moment nicht verfügbar oder wird nicht mehr hergestellt.
Schauen Sie bitte auf unsere Produktseite nach möglichen Alternativen
Beschreibung
Beschreibung
Suggested positive control: reticulocyte lysate.
Cell growth and viability is compromised by oxygen deprivation (hypoxia). Hypoxia-inducible factors, including HIF-1 alpha, HIF-1 beta (also designated Arnt 1), EPAS-1 (also designated HIF-2 alpha) and HIF-3 alpha, induce glycolysis, erythropoiesis and angiogenesis in order to restore oxygen homeostasis. Hypoxia-inducible factors are members of the Per-Arnt-Sim (PAS) domain transcription factor family. In response to hypoxia, HIF-1 alpha is upregulated and forms a heterodimer with Arnt 1 to form the HIF-1 complex. The HIF-1 complex recognizes and binds to the hypoxia responsive element (HRE) of hypoxia-inducible genes, thereby activating transcription.
Spezifikation
Spezifikation
HIF3A
Polyclonal
Unconjugated
Q9Y2N7, Q0VBL6
HIF3A
Bacterially expressed human HIF-3 alpha (N-terminus).
100 μL
Primary
Human, Mouse
Antibody
IgG
Western Blot
1 mg/mL
HIF3A
Hypoxia inducible factor 3a
Rabbit
Antigen affinity chromatography
RUO
53417, 64344
4° C, do not freeze