Learn More
P4HTM Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA530752
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: A549, H1299, HCT116. Predicted reactivity: Mouse (94%), Bovine (92%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
P4HTM Polyclonal antibody specifically detects P4HTM in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Spezifikation
P4HTM | |
Polyclonal | |
Unconjugated | |
P4HTM | |
4933406E20Rik; AI853847; BB128974; EGLN4; FLJ20262; HIFPH4; HIF-PH4; HIF-prolyl hydroxylase 4; HPH-4; hypoxia inducible factor prolyl 4 hydroxylase; hypoxia-inducible factor prolyl 4-hydroxylase; hypoxia-inducible factor prolyl hydroxylase 4; P4H TM; P4H with transmembrane domain; P4HTM; P4H-TM; PH 4; PH4; PH-4; PHD4; proline 4-hydroxylase; prolyl 4-hydroxylase, transmembrane; prolyl 4-hydroxylase, transmembrane (endoplasmic reticulum); Prolyl hydroxlase domain-containing 4; RGD1311848; Transmembrane prolyl 4-hydroxylase | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
54681 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
Q9NXG6 | |
P4HTM | |
Recombinant fragment corresponding to a region within amino acids 99 and 382 of Human HIF Prolyl Hydroxylase 4 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |