Learn More
PHD2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA116525
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
This antibody does not react with mouse samples. Suggested positive control: antigen standard for EGLN1 (transient overexpression lysate), human glioblastoma cells.
PHD2 Polyclonal antibody specifically detects PHD2 in Human, Non-human Primate, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunomicroscopy, Immunoprecipitation, Western Blot.Abbreviations: AP=Affinity Purification, ChIP=Chromatin IP, CM=Confocal Microscopy, DB=Dot Blot, EIA=ELISA, EM=Electron Microscopy, FC=Flow Cytometry, GS=Gel Supershift, IC=Immunocytochemistry, IF=Immunofluorescence, IH=Immunohistology/Immunohistochemistry, IP=Immunoprecipitation, IPK=IP Kinase Assay, KA=Kinase Assay, NEUT=Neutralization, PIA=Peptide Inhibition Assay, RIA=Radioimmunossay, TCA=T Cell Activation, WB=Western Blotting, Species: Am=Amphibian, Av=Avian, B=Bovine, Ca=Cat ,Ck=Chicken, D=Dog, Dr=Deer, Ds=Drosophila, El=Elk, Fg=Frog, Fs=Fish, Ft=Ferret, G=Goat, Gp=Guinea Pig, H=Horse, Hm=Hamster, Hu=Human, K=Kangaroo, Ms=Mouse, Nhp=Non-Human Primate, O=Ovine, P=Porcine, Rb=Rabbit, Rp=Reptile, Rt=Rat, Ys=Yeast, Xn=Xenopus, (wk)=cross reacts weakly
Spezifikation
PHD2 | |
Polyclonal | |
Unconjugated | |
Egln1 | |
AI503754; C1orf12; C1ORF12 antibody; ECYT3; egl nine homolog 1; egl nine-like protein 1; egl-9 family hypoxia inducible factor 1; egl-9 family hypoxia-inducible factor 1; EGLN 1 EGLN1; EGLN1; HALAH; HIF PH2; HIF prolyl hydroxylase 2; HIF-P4H 2; Hif-p4h-2; HIFPH2; HIF-PH2; HIF-prolyl hydroxylase 2; HPH2; HPH-2; Hypoxia-inducible factor prolyl hydroxylase 2; ORF13; P4H2; Phd2; PHD-2; PNAS-118; PNAS-137; Prolyl Hy; Prolyl hydroxylase domain-containing protein 2; SM20; SM-20; zinc finger MYND domain-containing protein 6; ZMYND6 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
308913, 54583 | |
4° C, do not freeze | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunomicroscopy, Immunoprecipitation, Western Blot | |
1 mg/mL | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
P59722, Q9GZT9 | |
Egln1 | |
A synthetic peptide corresponding to a C-terminal portion of human PHD2/HIF Prolyl Hydroxylase 2 (between amino acids 350-426) | |
100 μL | |
Primary | |
Human, Non-human Primate, Rat | |
Antibody | |
IgG |