Learn More
HIF1AN Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA534852
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: HCT116, MCF-7. Predicted reactivity: Mouse (98%), Rat (98%), Zebrafish (86%), Xenopus laevis (90%), Bovine (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
HIF1AN Polyclonal antibody specifically detects HIF1AN in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
HIF1AN | |
Polyclonal | |
Unconjugated | |
HIF1AN | |
2310046M24Rik; A830014H24Rik; DKFZp762F1811; factor inhibiting HIF1; factor inhibiting HIF-1; FIH; FIH 1; FIH1; FIH-1; FLJ20615; FLJ22027; Hif1an; hypoxia inducible factor 1 alpha subunit inhibitor; hypoxia inducible factor 1 subunit alpha inhibitor; hypoxia inducible factor 1, alpha subunit inhibitor; hypoxia-inducible factor 1, alpha subunit inhibitor; hypoxia-inducible factor 1-alpha inhibitor; Hypoxia-inducible factor asparagine hydroxylase; peptide-aspartate beta-dioxygenase | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55662 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q9NWT6 | |
HIF1AN | |
Recombinant fragment corresponding to a region within amino acids 78 and 349 of HIF1AN (Uniprot ID#Q9NWT6) | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |