missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HINT1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA520613
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
A suggested positive control is Jurkat cell lysate. PA5-20613 can be used with blocking peptide PEP-0733.
HINT1 Polyclonal antibody specifically detects HINT1 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Western Blot.Spezifikation
HINT1 | |
Polyclonal | |
Unconjugated | |
Hint1 | |
17 kDa inhibitor of protein kinase C; AA673479; Adenosine 5'-monophosphoramidase; FLJ30414; FLJ32340; HINT; Hint1; histidine triad nucleotide binding protein 1; histidine triad nucleotide-binding protein 1; Ipk1; NMAN; PKC inhibitor/ interacting protein; Pkci; PKCI1; PKCI-1; Prkci; PRKCNH1; protein kinase C inhibitor 1; protein kinase C, iota; protein kinase C-interacting protein 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
15254, 3094, 690660 | |
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
Liquid |
Immunocytochemistry, Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
P49773, P62959, P70349 | |
Hint1 | |
A 17 amino acid peptide near the center of the human HINT1. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |