missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZDHHC17 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marke: Invitrogen PA518336
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
This antibody is predicted to react with bovine, canine and rat based on sequence homology.
ZDHHC17 Polyclonal antibody specifically detects ZDHHC17 in Human, Mouse samples. It is validated for Immunohistochemistry, Western Blot.Spezifikation
ZDHHC17 | |
Polyclonal | |
Unconjugated | |
ZDHHC17 | |
A230053P19Rik; BB187739; D130071N24Rik; DHHC-17; Hip14; HIP-14; HIP3; HIP-3; HSPC294; huntingtin interacting protein 14; huntingtin interacting protein 3; Huntingtin interacting protein H; huntingtin yeast partner H; Huntingtin-interacting protein 14; huntingtin-interacting protein 3; Huntingtin-interacting protein H; HYPH; Kiaa0946; membrane-associated DHHC17 zinc finger protein; palmito; palmitoyltransferase ZDHHC17; Putative MAPK-activating protein PM11; putative NF-kappa-B-activating protein 205; ZDHHC17; Zinc finger DHHC domain-containing protein 17; zinc finger DHHC-type containing 17; zinc finger, DHHC domain containing 17; zinc finger, DHHC-type containing 17 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
23390, 320150 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q80TN5, Q8IUH5 | |
ZDHHC17 | |
Synthetic peptide sequence (CYDQISGSGYQLV) corresponding to the C-terminus amino acids of ZDHHC17 | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |